Would you like to log in again?
Interleukin-8/NAP-1, IL-8, Human ELISA, BioVendor
Supplier: BioVendor
RAF074R
102552-398EA 1054.94 USD InStock
102552-398
Interleukin-8/NAP-1, IL-8, Human ELISA, BioVendor
Assays ELISAs
IL-8 (8 kDa, 72 amino acids) is a non-glycosylated protein and it is produced by the processing of a precursor protein of 99 amino acids.
  • Calibration Range: 15.6-1000 pg/mL
  • Limit of Detection: 2 pg/mL
  • Cell culture supernatant, Plasma-EDTA, Plasma-Heparin, Serum: 50 uL/well
IL-8 (8 kDa, 72 amino acids) is a non-glycosylated protein and it is produced by the processing of a precursor protein of 99 amino acids. IL-8 is present in the synovial fluids of patients with rheumatoid arthritis and has been purified from plaque psoriasis scales. It is associated with sepsis, asthma and glomerulonephritis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty