Would you like to log in again?
IGFBP-1 Human ELISA, BioVendor
Supplier: BioVendor
RCD018R
102552-422EA 610.74 USD InStock
102552-422
IGFBP-1 Human ELISA, BioVendor
Assays ELISAs
Insulin-like growth factor binding protein-1 (IGFBP-1) is one of six proteins that specifically bind insulin-like growth factors I and II (IGF-I and IGF-II) in body fluids and tissues.
  • Calibration Range: 1-250 ug/mL
  • Limit of Detection: 0.5 ug/mL
  • Serum: 25 uL/well
Insulin-like growth factor binding protein-1 (IGFBP-1) is one of six proteins that specifically bind insulin-like growth factors I and II (IGF-I and IGF-II) in body fluids and tissues. IFGBP-1 contains 234 amino acids, with a predicted molecular mass of 25 kDa. The major sites of IGFBP-1 synthesis are the fetal /adult liver and decidualized endometrium. Serum levels of IGFBP-1, which reflect its synthesis by the liver, exhibit considerable diurnal variation. Circulating IGFBP-1 levels are highest early in the morning and lowest in the evening. The levels are high in the fetus and newborn, but decline steadily until puberty. The mean level of IGFBP-1 in healthy adults is 4.4 μg/L (range 0.6–14.4 μg/L). After about 65 years of age, serum IGFBP-1 levels begin to increase. There is also an inverse correlation between body mass index (BMI) and fasting serum IGFBP-1 concentrations. The most important regulator of circulating IGFBP-1 is insulin. Fasting insulin and IGFBP-1 concentrations are inversely correlated. During a 3-h glucose tolerance test, there is a decrease of about 50% in serum IFGBP-1 levels. Eating a meal also has a decreasing effect. In insulin-dependent diabetes (IDDM), serum IGFBP-1 levels are elevated. In non-insulin dependent diabetes, in which insulin levels are high, serum IGFBP-1 is decreased. Low levels of IGFBP-1 have also been observed in the following cases: acromegaly, Cushing’s syndrome and polysystic ovarian syndrome (PCO).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty