Would you like to log in again?
IP-10 Mouse ELISA, BioVendor
Supplier: BioVendor
RAF077R
102552-444EA 1014.71 USD InStock
102552-444
IP-10 Mouse ELISA, BioVendor
Assays ELISAs
IP-10 (Interferon-gamma inducible Protein 10kDa) also known as CXCL10, is secreted by several cell types in response to IFNg and LPS.
  • Calibration Range: 7.8-500 pg/mL
  • Limit of Detection: 6.5 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-EDTA, Serum: 50 uL/well
IP-10 (Interferon-gamma inducible Protein 10kDa) also known as CXCL10, is secreted by several cell types in response to IFNg and LPS. These cell types include monocytes, endothelial cells and fibroblasts.The gene for IP-10 is located on chromosome 4 in a cluster among several other cytokines and encodes a 98 amino acid precursor protein. IP-10 has been attributed to several roles, such as chemoattraction for monocytes and T cells (but not for neutrophils), inhibition of bone marrow colony formation and angiogenesis, promotion of T cells adhesion molecule expression. IP-10 shares a common receptor, CXCR3, with the chemokine MIG, but has been shown to play a distinct role in host defense in infections. IP-10 expression has been associated with HIV infection, is involved in inflammatory skin disease (6) and other allergic diseases; it appears in inflammation of the nervous system and in Alzheimer’s disease (astrocytes expressing IP-10 are commonly associated with senile plaques).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty