Would you like to log in again?
LIF Human ELISA, BioVendor
Supplier: BioVendor
RAF078R
102552-456EA 1054.98 USD InStock
102552-456
LIF Human ELISA, BioVendor
Assays ELISAs
LIF is a bioactive protein intended for use in cell culture applications.
  • Calibration Range: 3.2-200 pg/mL
  • Limit of Detection: 0.66 pg/mL
  • Cell culture supernatant, Plasma, Serum: 50 uL/well
Cytokines and chemokines and related molecules, Oncology
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty