Would you like to log in again?
sL-Selectin Human ELISA, BioVendor
Supplier: BioVendor
RAF108R
102552-466EA 1054.98 USD InStock
102552-466
sL-Selectin Human ELISA, BioVendor
Assays ELISAs
L-selectin could provide more detailed insights into the pathological modifications during various diseases.
  • Calibration Range: 0.39-25 ng/mL
  • Limit of Detection: 0.19 ng/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum, Urine: 50 µl (1:100 prediluted)
Determination of soluble/circulating L-selectin could provide more detailed insights into the pathological modifications during various diseases.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty