Would you like to log in again?
Monocyte Chemotactic Protein-1, MCP-1, Human ELISA, BioVendor
Supplier: BioVendor
RAF081R
102552-478EA 1054.98 USD InStock
102552-478
Monocyte Chemotactic Protein-1, MCP-1, Human ELISA, BioVendor
Assays ELISAs
Monocyte chemotactic protein-1 (MCP-1) is a member of the C-C family of chemokines.
  • Calibration Range: 15.6-1000 pg/mL
  • Limit of Detection: 2.2 pg/mL
  • Cell culture supernatant, Plasma-EDTA, Plasma-Heparin, Serum: 20 uL/well
Monocyte chemotactic protein-1 (MCP-1) is a member of the C-C family of chemokines. Chemokines are mediators that share the capacity to induce directional migration of leukocytes. MCP-1 is a chemoattractant for many cell types such as monocytes, neutrophils, hepatic stellate cells, granulocytes and T lymphocytes. MCP-1 is believed to play a role in a variety of pathological conditions, including inflammatory and immune reactions, atherosclerosis and neoplasia. Monocyte Chemotactic Protein-1 (MCP-1) is also known as Monocyte Chemotactic Activating Factor (MCAF) or CCL2.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty