Would you like to log in again?
Matrix Metalloproteinase-9, MMP-9, Human ELISA, BioVendor
Supplier: BioVendor
RAF079R
102552-486EA 1054.98 USD InStock
102552-486
Matrix Metalloproteinase-9, MMP-9, Human ELISA, BioVendor
Assays ELISAs
Matrix metalloproteinases (MMPs) form a family of enzymes with major actions in the remodelling of extracellular matrix (ECM) components.
  • Calibration Range: 0.23-15 pg/mL
  • Limit of Detection: 0.05 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-Heparin, Serum: 10 uL/well (1:25 prediluted)
Matrix metalloproteinases (MMPs) form a family of enzymes with major actions in the remodelling of extracellular matrix (ECM) components. MMP-9, also called Gelatinase B, is the most complex family member in terms of domain structure and regulation of its activity. MMP-9 activity is under strict control at various levels: transcription of the gene by cytokines and cellular interactions; activation of the pro-enzyme by a cascade of enzymes comprising serine proteases and other MMPs; and regulation by specific tissue inhibitors of MMPs (TIMPs) or unspecific inhibitors. Further glycosylation has a limited effect on the net activity of Gelatinase B and chemotactic factors are another level of control of activity. The main function of MMPs is degradation of the extracellular physiologic function including wound healing, bone resorption and mammary involution. MMPs also contribute to pathological conditions such as rheumatoid arthritis, coronary artery disease and cancer. They are thought to promote the growth of these tumor cells once they have metastasized. MMP-9 plays an important role in tumor invasion, angiogenesis and metastasis. Increased levels of MMP-9 have been demonstrated in colorectal cancer, in acute leukemia, breast cancer, human melanoma and bladder cancer. MMP-9 potentiates pulmonary metastasis formation and glioma invasion. Apart from its function in tumor formation and spread, MMP-9 with its destructive effects is in close association with lung diseases and asthma. Leukocyte recruitment to the central nervous system MMP-9 means an involvement in the pathogenesis of multiple sclerosis. MMP-9 has a crucial role in reproductive endocrinology and shows a changed expression pattern after myocardial infarction.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty