Would you like to log in again?
Osteocalcin, OST, Human ELISA, BioVendor
Supplier: BioVendor
RIS002R
102552-498EA 689.05 USD InStock
102552-498
Osteocalcin, OST, Human ELISA, BioVendor
Assays ELISAs
Osteocalcin or bone Gla protein (B.G.P) is the major non-collagen protein of the bone matrix.
  • Calibration Range: 2–50 ng/mL
  • Limit of Detection: 0.4 ng/mL
  • Serum: 25 uL/well
Osteocalcin or bone Gla protein (B.G.P) is the major non-collagen protein of the bone matrix. It has a molecular weight of 5800 Da and contains 49 amino-acids, including 3 residues of gamma carboxyl glutamic acid. Osteocalcin is synthesized in the bone by the osteoblasts. After production, it is partly incorporated in the bone matrix and the rest is found in the blood circulation. The exact physiological function of osteocalcin is still unclear. A large number of studies show that the circulating levels of osteocalcin reflect the rate of bone formation.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty