Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pepsinogen I Human ELISA, BioVendor

Supplier: BioVendor
RIS007R
102552-512EA 1023.58 USD InStock
102552-512
Pepsinogen I Human ELISA, BioVendor
Assays ELISAs
Pepsinogen consists of a single polypeptide chain of 375 amino acids with an average molecular weight of 42 kDa.

  • Calibration Range: 3–300 ng/mL
  • Limit of Detection: 0.5 ng/mL
  • Serum: 25 uL/well

Pepsinogen consists of a single polypeptide chain of 375 amino acids with an average molecular weight of 42 kDa. Pepsinogen I is synthesized at gastric chief cells and mucous neck cells, while pepsinogen II is produced not only by gastric chief cells, mucous neck cells, but also by clear mucous cells of antrum, etc. The clinical applications of measuring pepsinogen I and pepsinogen II are of useful aid in diagnosing severe atrophic gastritis and stomach cancer. It was suggested that the measurement of serum pepsinogens served as a “serological biopsy” for predicting the presence of atrophic gastritis or superficial gastritis. Atrophic Gastritis: It was found that a serum pepsinogen I level below 20 ng/ml was highly specific for severe atrophic gastritis. It is also observed that serum pepsinogen I levels fell with increasing severity of mucosal damage in atrophic gastritis. The diagnostic sensitivity and specificity of serum pepsinogen I level for advanced atrophic corpus gastritis are about 92% and 90% respectively. On the other hand, the decrease in serum pepsinogen I levels in patients with pernicious anemia and atrophic gastritis was found to be associated with normal or raised pepsinogen II levels. Therefore, a pepsinogen I/pepsinogen II ratio is significantly lower than those with superficial gastritis or normal remnant mucosa. Stomach Cancer: Low serum pepsinogen I levels were found in patients with gastric cancer, with a threefold higher incidence. Other studies have concluded that low serum pepsinogen I levels may identify persons at increased risk for intestinal types of stomach cancer. Duodenal Ulcer: A low serum pepsinogen I level can exclude a diagnosis of duodenal ulcer. Although a high pepsinogen I level has less clinical useful for establishing the diagnosis of a duodenal ulcer, the combination of of hypergastrinemia and a highly elevated serum pepsinogen.

Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty