Would you like to log in again?
Phosphorylated Neurofilament H Human ELISA, Biovendor
Supplier: BioVendor
RD191138300R
102552-520EA 870.92 USD InStock
102552-520
Phosphorylated Neurofilament H Human ELISA, Biovendor
Assays ELISAs
96 wells (1 kit) - ELISA is a sandwich enzyme immunoassay for the quantitative measurement of pNF-H.
In the BioVendor Human Phosphorylated Neurofilament H ELISA, standards, quality controls and samples are incubated in microplate wells pre-coated with chicken polyclonal anti-pNF-H antibody. After 60 minutes incubation and washing, detection rabbit polyclonal anti-pNF-H antibody is added and incubated with captured pNF-H for 60 min. After another washing, HRP conjugated antibody against rabbit antibody is added. After 60 min incubation and the last washing step, the remaining conjugate is allowed to react with the substrate solution (TMB). The reaction is stopped by addition of acidic solution and absorbance of the resulting yellow product is measured. The absorbance is proportional to the concentration of pNF-H. A standard curve is constructed by plotting absorbance values against concentrations of standards, and concentrations of unknown samples are determined using this standard curve.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty