Would you like to log in again?
Placental Lactogen Human ELISA, BioVendor
Supplier: BioVendor
RIS0013R
102552-524EA 658.35 USD InStock
102552-524
Placental Lactogen Human ELISA, BioVendor
Assays ELISAs
HPL may become a regulator of feto-placental growth and other physiological alterations during pregnancy.
  • Calibration Range: 1.25–20 ng/mL
  • Limit of Detection: 0.3 ng/mL
  • Serum: 10 uL/well
The physiological role of human placental lactogen (HPL) has not yet been established, but the great similarity to human growth hormone has stimulated the hypothesis about a function as a regulator of feto-placental growth and other physiological alterations during pregnancy. It has been suggested that the maternal serumlevel may reflect on “index of placental function”. Depressed levels of HPL are seen in association with intrauterine death, fetal distress in labor and birth asphyxia. This association is particularly strong if depressed levels are seen repeatedly, implying a chronic state of placental and therefore fetal compromise. Depressed levels are usually not present if the pregnancy has proceeded uneventfully to term. Elevated levels of HPL are usually indicative of optimal pregnancy outcome in singleton pregnancies. However, high levels may indicate substantial fetal pathology in specific diseases, namely diabetes mellitus and fetal macrosomia, rhesus isoimmunization and hydrops fetalis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty