Would you like to log in again?
Pregnenolone Human ELISA, BioVendor
Supplier: BioVendor
RCD022R
102552-526EA 598.42 USD InStock
102552-526
Pregnenolone Human ELISA, BioVendor
Assays ELISAs
Pregnenolone is the first steroid to be derived from cholesterol in the pathway of steroidogenesis, and it is the common precursor for all of the adrenal and gonadal steroids.
  • Calibration Range: 0.1-25 ng/mL
  • Limit of Detection: 0.05 ng/mL
  • Serum: 50 uL/well
Pregnenolone is the first steroid to be derived from cholesterol in the pathway of steroidogenesis, and it is the common precursor for all of the adrenal and gonadal steroids. Its production occurs in the mitochondrion by cleavage of the C-20 side chain of cholesterol by the P-450SCC enzyme. Elevated pregnenolone levels occur in forms of congenital adrenal hyperplasia. The determination of pregnenolone in serum may be useful for studying its metabolite, pregnenolone sulfate, which has been reported to have various effects in the mammalian brain and central nervous system.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty