Would you like to log in again?
sPSGL-1 Human ELISA, BioVendor
Supplier: BioVendor
RAF113R
102552-544EA 1054.98 USD InStock
102552-544
sPSGL-1 Human ELISA, BioVendor
Assays ELISAs
Regulation in many physiological and pathological processes can take place with soluble PSGL-1.
  • Calibration Range: 1.6-50 U/mL
  • Limit of Detection: 0.99 U/mL
  • Cell culture supernatant, Serum: 100 µL (1:20 prediluted)
The family of selectins consists of three structurally and functionally related molecules. Lselectin is constitutively expressed on neutrophils, P-selectin is found on platelets and is stored in Weibel-Palade bodies from where it is transported to the cell surface upon endothelial activation. E-selectin is expressed on endothelial cells. Due to a common structural element, the amino-terminal lectin-like domain, the selectins are able to bind to carbohydrate ligands. Different putative ligand structures have been identified for which the selectins show high affinity. These structures include oligosaccharides, phophorylated saccharides, sulfopolysaccharides and lipids. It was shown that glycoproteins represent the biological relevant ligands for selectins. While the ligands for E- and L-selectins with primary binding activity have not been identified so far, the functionally most important ligand for P-selectin has been identified. The mucin-like glycoprotein PSGL-1 (P-selectin Glycoprotein Ligand-1) has been cloned and sequenced. PSGL-1 has been shown to be a transmembrane protein which forms homodimers via disulfide bridges of two 120 kDa chains. PSGL-1 is expressed on cells of myeloid, lymphoid and dendritic lineage. The binding of Pselectin is regulated by different degrees and forms of glycosylation. An interaction of Lselectin with PSGL-1 in the process of neutrophil aggregation has been shown. However, PSGL-1 does not seem to be the primary ligand for L-selectin. Presently it is not known which cells, apart from leukocytes, express PSGL-1 and what role PSGL-1 plays on these cells. The metastic potential of the majority of cells which bind to Pselectin is however in close correlation with the functional expression of PSGL-1 on these cells. The regulation of PSGL-1 is not yet well described. Glycosyltransferases sure play an important role in activation. The deactivation of PSGL-1 is so far unclear. The cleavage of the protein from the cell surface is one mechanism involved in the deactivation process. Following this shedding, a soluble form of PSGL-1 is detectable in the circulation. This soluble isoform of PSGL-1 is still capable of binding to P-selectin, thus representing a competitor for cellular PSGL-1 through which regulation in many physiological and pathological processes can take place.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty