Would you like to log in again?
Secretagogin Human ELISA, BioVendor
Supplier: BioVendor
RD191120200R
102552-554EA 929.25 USD InStock
102552-554
Secretagogin Human ELISA, BioVendor
Assays ELISAs
Secretagogin may be involved in suppressing cell growth, and it has been proposed as a novel biomarker for diagnosis of different forms of cancer.
  • Calibration Range: 62.5-2000 pg/mL
  • Limit of Detection: 11 pg/mL
  • Plasma-EDTA, Serum: 50 uL/well
Secretagogin (SCGN, SEGN, CALBL, setagin) is a Ca-binding protein which consists of six-EF hand Ca-binding domains and is characterised by molecular weight of 32 kDa. Generally, Ca-binding proteins seem to be involved in several pathological conditions in the CNS, such as epilepsy or neurodegenerative disorders, for example Alzheimer´s disease. Secretagogin is expressed in central nervous system, particularly in the cerebellum, pituitary gland and hypotalamus, but also in thalamic tissue. Moreover, recent study shown, that secretagogin is detectable in human serum after ischemic brain damage. The secretagogin mRNA was identified as one of the most abundant transcripts in pancreatic islets, highlighting the potential importance of secretagogin in this tissue. Secretagogin is down-regulated in pancreatic islets exposed to high concentration of glucose, in human non-functional pituitary adenomas and in adenocarcinomas. These results suggest that secretagogin may be involved in suppressing cell growth, and it has been proposed as a novel biomarker for diagnosis of different forms of cancer. Additionally, its strong expression in insulin secreting cells, its influence on insulin expression might contribute to insulin homeostasis. Areas of investigation: Neurodegenerative disease, Brain injury, Insulin secretion and Cancer.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty