Would you like to log in again?
Sperm-Antibody Human ELISA, BioVendor
Supplier: BioVendor
RIS0017R
102552-558EA 724.34 USD InStock
102552-558
Sperm-Antibody Human ELISA, BioVendor
Assays ELISAs
Recommended for the diagnosis of immunologically caused disorders of fertility.
  • Calibration Range: 62–500 U/mL
  • Serum: 5 uL/well
Antibodies directed against spermatozoa antigens may cause infertility in women or men. The application of the Anti-Spermatozoa Antibody ELISA from Biovendor is recommended for the diagnosis of immunologically caused disorders of fertility. Unwanted childlessness is a growing problem with which up to 20% of all couples in the reproductive age are confronted temporarily or long-term. In 20% of these cases the presence of anti-spermatozoa antibodies in the male or the female patient is detectable.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty