Would you like to log in again?
Testosterone, Free Human ELISA, BioVendor
Supplier: BioVendor
RCD016R
102552-576EA 589.62 USD InStock
102552-576
Testosterone, Free Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of testosterone.
  • Calibration Range: 0.25-125 pg/mL
  • Limit of Detection: 0.17 pg/mL
  • Serum: 25 uL/well
For the direct quantitative determination of free testosterone by enzyme immunoassay in human serum.
Testosterone is a C-19 steroid secreted from the testis and the adrenal cortex in men and from the adrenal cortex and ovaries in women. Testosterone is also produced by peripheral tissues from androstenedione, which is of little physiological significance in men, however in women about half of the circulating testosterone is derived from this origin. Testosterone measurements are used mainly for clinical evaluation of hypogonadism in males and hyperandrogenic states in females.
The principle of the following enzyme immunoassay test follows the typical competitive binding scenario. Competition occurs between an unlabeled antigen (present in standards,controls and patient samples) and an enzyme-labelled antigen (conjugate) for a limited number of antibody binding sites on the microwell plate. The washing and decanting procedures remove unbound materials. After the washing step, the enzyme substrate is added. The enzymatic reaction is terminated by addition of the stopping solution. The absorbance is measured on a microtiter plate reader. The intensity of the color formed is inversely proportional to the concentration of free testosterone in the sample. A set of standards is used to plot a standard curve from which the amount of free testosterone in patient samples and controls can be directly read. The BIOVENDOR free testosterone kit utilizes a highly specific rabbit anti-testosterone polyclonal antibody at a low binding capacity (Keq x concentration) to keep minimum disturbances of the testosterone-protein equilibrium. The other components in the test system are also optimized in order to not alter the original free testosterone concentration.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty