Would you like to log in again?
Free Thyroxine, fT4, Human ELISA, BioVendor
Supplier: BioVendor
RCD015R
102552-594EA 540.31 USD InStock
102552-594
Free Thyroxine, fT4, Human ELISA, BioVendor
Assays ELISAs
The free T4 assay is a one-step competitive ELISA system that is rapid and easy to perform compared to equilibrium dialysis and ultrafiltration methods.
  • Calibration Range: 2-95 pg/mL
  • Limit of Detection: 1 pg/mL
  • Serum: 25 uL/well
Thyroxine (T4), the principal thyroid hormone, circulates in blood almost completely bound to carrier proteins. However, only the free (unbound) fraction of thyroxine is considered to be biologically active. The main carriers of thyroxine are thyroxine-binding globulin (TBG), pre-albumin and albumin. The measurement of free thyroxine (fT4) levels correlate better with the clinical status than total thyroxine levels. The free T4 assay is a one-step competitive ELISA system that is rapid and easy to perform compared to equilibrium dialysis and ultrafiltration methods, which are cumbersome and time-consuming. This system employs a highly specific monoclonal antibody and a non-analog tracer that was proved experimentally to have no significant binding to TBG and albumin. In the euthyroid, normal population the free T4 concentration is 7 – 22 pg/ml. The level of free T4 is decreased in hypothyroidism while in thyrotoxic patients the level of free T4 is increased. This assay is used at times with other thyroid tests for in vitro diagnostic purposes and for assessing patients who are receiving thyroid treatments.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty