Would you like to log in again?
TIE-2 Human ELISA, BioVendor
Supplier: BioVendor
RAF126R
102552-598EA 1055.11 USD InStock
102552-598
TIE-2 Human ELISA, BioVendor
Assays ELISAs
TIE-2 maintains a long-term, quiescent population of hematopoietc stems cells in the bone marrow.
  • Calibration Range: 0.16-10 ng/mL
  • Limit of Detection: 0.03 ng/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 50 µL/well
The tyrosine kinase receptor TIE-2 was initially identified as a specific endothelial growth factor receptor that mediated several properties of endothelial cells under both physiologcal and pathological conditions. Angiopoietins are the natural ligands of TIE-2 and they induce TIE-2– depending signalling, including survival and apoptosis of endothelial cells, vascular permeability and regulate capillary sprouting. In general, the outcome of tie receptor signalling depends on which vascular bed is involved, and crosstalk between different VEGFs has an important modulating effect on the properties of the ligands. TIE-2 plays an important role in several vascular diseases, known as vascular malformation (e.g. venous malformation). Independent of angiogenesis and its involvement in lymphangiogenesis, TIE-2 maintains a long-term, quiescent population of hematopoietc stems cells in the bone marrow. In a tumor model, a subset of monocytes was found to be positve for TIE-2 and that they have an important function in paracrine support of nascent blood vessels. During cancer formation and spreading, TIE-2 was found to be overexpressed in tumor vessels. However, outsite the vascular compartment TIE-2 is expressed in several types of cancer including leukemia and in gastric tumors, breast tumors and gliomas. In gliomas, TIE-2 expression in the neoplastic glial cells was significantly associated with progression from a lower to a higher grade where it seems to regulate glioma cell adhesion to extracellular matrix. Together with the reported fact that malignant gliomas epress high levels of Ang1, suggest the existence of an autocrine loop for cell matrix interaction. The possible role of TIE-2 in tumors is more currently under investigation. Because it is expressed in several cellular lineages insite the tumor, the receptor may be an attractive target for cancer therapy. A natural occuring soluble TIE-2 receptor fragment of 75 kDa (sTIE-2) is generated by shedding. Soluble TIE-2 inhibits angiopoietin-mediated TIE-2 phosphorylation and anti-apoptosis. TIE-2 shedding is mediated by PI2K/Akt and p38 MAPK. The serum levels of sTIE-2 in patients is in the lower ng/ml range and dependent of the disease. Soluble TIE-2 was also measured in preterm infants including the ROP syndrome (retinopathy of prematurity). Elevated plasma levels in active ROP patients were observed for sTIE-2. In experimental tumor mouse models sTIE-2 can be used for tumor regression and prolonged the tumor-free survival in 80% of the animals.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty