Would you like to log in again?
TNF alpha Human ELISA, BioVendor
Supplier: BioVendor
RAF128R
102552-602EA 1055.05 USD InStock
102552-602
TNF alpha Human ELISA, BioVendor
Assays ELISAs
Tumor Necrosis Factor alpha (TNF alpha), also known as cachectin, is a polypeptide cytokine produced by monocytes and macrophages.
  • Calibration Range: 7.8-500 pg/mL
  • Limit of Detection: 2.3 pg/mL
  • Body fluids, Culture medium, Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum, Urine: 50 µL/well
Tumor Necrosis Factor alpha (TNF alpha), also known as cachectin, is a polypeptide cytokine produced by monocytes and macrophages. It functions as a multipotent modulator of immune response and further acts as a potent pyrogen. TNF alpha circulates throughout the body responding to stimuli (infectious agents or tissue injury), activating neutrophils, altering the properties of vascular endothelial cells, regulating metabolic activities of other tissues, as well as exhibiting tumoricidal activity by inducing localized blood clotting. TNF alpha also inhibits lipoprotein lipase activity resulting in cachexia, a physical wasting condition. Activation of B-cells by the Epstein Barr virus can be inhibited by TNF alpha. Due to its varied actions throughout the immune system, TNF alpha may play a role in the pathogenesis of many disease states. TNF alpha production is mediated by the action of lymphokines and endotoxins on the macrophage. Purified monocytes produce TNF alpha within four hours of stimulation by recombinant IL-2 and there is some in vitro evidence to suggest that TNF alpha is expressed at high levels and with prolonged kinetics in T cells stimulated by both CD2 and CD28. Secretion of TNF alpha is enhanced by gamma interferon. TNF then induces or enhances the specific production of Class I MHC antigen, GM-CSF, and IL-1. Recent evidence has suggested an intracellular role for this peptide. TNF alpha may play a significant role in the pathogenesis of inflammatory disease of the joints and other tissues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty