Would you like to log in again?
TNF alpha Total Monkey ELISA, BioVendor
Supplier: BioVendor
RAF131R
102552-612EA 1015.54 USD InStock
102552-612
TNF alpha Total Monkey ELISA, BioVendor
Assays ELISAs
TNF-α (17.5 kDa, 157 amino acid) is a non-glycosylated protein that is a potent lymphoid factor, which exerts cytotoxic effects on a wide range of tumor cells and other target cells.
  • Calibration Range: 28.1-1800 U/mL
  • Limit of Detection: 6 U/mL
  • Baboon serum, Cell culture supernatant, Cynomolgus serum: 50 µL/well
TNF-α (17.5 kDa, 157 amino acid) is a non-glycosylated protein that is a potent lymphoid factor, which exerts cytotoxic effects on a wide range of tumor cells and other target cells. It is secreted by macrophages, monocytes, neutrophils, T-cells and NK-cells following their stimulation by bacterial lipopolysaccharides. TNF-α has been suggested to play a pro-inflammatory role and has been detected in synovial fluid of patients with rheumatoid arthritis.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty