Would you like to log in again?
sTNF-R, 80 kDa, Human ELISA, BioVendor
Supplier: BioVendor
RAF115R
102552-620EA 1055.11 USD InStock
102552-620
sTNF-R, 80 kDa, Human ELISA, BioVendor
Assays ELISAs
This assay will help to clarify the possible diagnostic and prognostic value of circulating sTNF-R (60 kDa) in various neoplastic and inflammatory diseases.
  • Calibration Range: 0.16-10 ng/mL
  • Limit of Detection: 0.1 ng/mL
  • Cell culture supernatant, Plasma, Serum, Urine: 10 µL/well
Tumor Necrosis Factor (TNF) was originally discovered in sera of animals and was found to cause hemorrhagic necrosis of some transplantable mouse and human tumors and to exhibit primarily cytotoxic activities against tumor but not normal cells in vitro. The TNF family consists of two proteins designated TNFα, also called cachectin, and TNFβ, also called lymphotoxin, which are pleiotropic cytokines that can mediate a wide variety of biological effects.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty