Would you like to log in again?
TRAIL Human ELISA, BioVendor
Supplier: BioVendor
RAF135R
102552-626EA 1054.98 USD InStock
102552-626
TRAIL Human ELISA, BioVendor
Assays ELISAs
TRAIL is involved in lymphatic neoplastic disorders and thyroid diseases and in the treatment of cancer like melanoma.
  • Calibration Range: 15.6-1000 pg/mL
  • Limit of Detection: 5 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 50 µL/well
TRAIL is a member of the TNF family which mediates cell death in a wide variety of malignant cell lines and primary tumor cells. It induces two different signals: cell death mediated by caspases and gene induction mediated by NFkappaB. TRAIL, which is ubiquitously expressed, exhibits very complex signalling and tumor-selective induction of apoptosis. TRAIL is involved in lymphatic neoplastic disorders and thyroid diseases and in the treatment of cancer like melanoma.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty