Would you like to log in again?
Triiodothyronine, Total, T3, Human ELISA, BioVendor
Supplier: BioVendor
RCD025R
102552-644EA 524.45 USD InStock
102552-644
Triiodothyronine, Total, T3, Human ELISA, BioVendor
Assays ELISAs
T3 has an influence on oxygen consumption and heat production in virtually all tissues.
  • Calibration Range: 0.2-10 ng/mL
  • Limit of Detection: 0.16 ng/mL
  • Serum: 50 uL/well
Triiodothyronine (T3) and thyroxine (T4) are the two active thyroid hormones found in the blood stream. About 80% of T3 is produced by the deiodination of T4 in the peripheral tissue and the other 20% is produced directly from the thyroid gland. T3 is transported through the peripheral blood stream bound to serum proteins, namely thyroxine binding globulin, thyroid binding prealbumin and albumin. About 0.3% of the total T3 is unbound and is therefore considered the free fraction. T3 has an influence on oxygen consumption and heat production in virtually all tissues. The hormone also plays a critical role in growth, development and sexual maturation of growing organisms. T3 is one parameter used in the clinical diagnosis and differentiation of thyroid disease, particularly hyperthyroidism. In most hyperthyroid patients, both serum T3 and serum T4 levels are elevated. Serum T3 levels are a sensitive indicator of the impending hyperthyroid state often preceeding elevated T4 and free thyroxine index values. Serum T3 levels are clinically significant in both the diagnosis of thyroid disease and in the detection of T3-thyrotoxicosis. However, it has been demonstrated that T3 levels may be affected by a number of medications, acute and chronic stress, and a variety of acute and chronic non-thyroidal illnesses. It is therefore necessary to differentiate those results that are due to thyroid dysfunction from those related to non-thyroidal diseases.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty