Would you like to log in again?
sVAP-1 Human ELISA, BioVendor
Supplier: BioVendor
RAF117R
102552-650EA 1054.98 USD InStock
102552-650
sVAP-1 Human ELISA, BioVendor
Assays ELISAs
Vascular adhesion protein-1 (VAP-1) is one of the endothelial cell adhesion molecules that mediates lymphocyte binding to the endothelium under shear conditions.
  • Calibration Range: 31.3-2000 pg/mL
  • Limit of Detection: 19 pg/mL
  • Amniotic fluid, Cell culture supernatant, Plasma-EDTA, Plasma-Heparin, Serum: 10 µL (1:100 prediluted)
The ability of leukocytes to extravasate from the vasculature to the tissue space is a fundamental feature of the normal immune system. Several cell adhesion molecules play a key role in this complex process of initial and stable adhesion and diapedesis of leukocytes across the endothelial barrier. Vascular adhesion protein-1 (VAP-1) is one of the endothelial cell adhesion molecules that mediates lymphocyte binding to the endothelium under shear conditions. It is constitutively expressed mainly in high endothelial venules in peripheral lymph nodes. The expression of VAP-1 is induced by chronic inflammation in the vessels of the tonsil, gut, skin and synovium. VAP-1 is also present on sinusoidal and vascular endothelium in the liver under both normal and inflammatory conditions, however it is not found on any of the blood leukocytes. VAP-1 expression has furthermore been shown in human cervix and vagina mucosa. In psoriasis and allergic lesions, VAP-1 is markedly upregulated. The mature VAP-1 molecule is a 170 kDa homodimeric glycoprotein that consists of two 90 kDa subunits held together by disulfide bonds. VAP-1 has a large extracellular domain, a single-pass transmembrane domain, and a short cytoplasmic tail. The molecule has abundant sialic acid decorations that are essential to its adhesive function, because VAP-1 is unable to mediate lymphocyte adhesion to desialylated vessels. The leukocyte ligand for VAP-1 is currently unknown. Induction of VAP-1 has been shown at sites of inflam-mation, such as in inflammatory bowel diseases and chronic dermatoses, where expression of VAP-1 is clearly increased. It is constitutively expressed on hepatic endothelium playing a critical role in regulation of T-cell recirculation to the liver. Strong expression of VAP-1 on tumor endothelium distinguishes human hepatocellular carcinomas from colorectal hepatic metastases. A circulating form of human Vascular Adhesion Protein-1 (sVAP-1) has been characterized. This sVAP-1 has been shown to be elevated in sera of patients with certain liver diseases and a correlation with the diagnosis of the patients was demonstrated.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty