Would you like to log in again?
VEGF-A Human ELISA, BioVendor
Supplier: BioVendor
RAF136R
102552-660EA 1054.98 USD InStock
102552-660
VEGF-A Human ELISA, BioVendor
Assays ELISAs
96 wells (1 kit) - ELISA is a competitive enzyme immunoassay for the quantitative measurement of VEGF-A.
  • Calibration Range: 15.6-1000 pg/mL
  • Limit of Detection: 7.9 pg/mL
  • Cell culture supernatant, Plasma-Citrate, Plasma-EDTA, Plasma-Heparin, Serum: 50 µL/well
The Human VEGF-A ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of Human VEGF-A.
Normal tissue function depends on a regular supply of oxygen through the blood vessels. Understanding the formation of blood vessels has become the focus of a major research effort throughout the last decade. Vasculogenesis in the embryo is the process by which new blood vessels are generated de novo from primitive precursor cells. Angiogenesis is the process of new blood vessel formation from pre-existing vasculatures. It plays an essential role in development, normal tissue growth, wound healing, the female reproductive cycle (placental development, ovulation, corpus luteum) and also plays a major role in various diseases.
An anti-human VEGF-A coating antibody is adsorbed onto microwells. Human VEGF-A present in the sample or standard binds to antibodies adsorbed to the microwells. First Incubation - Following incubation unbound biological components are removed during a wash step. A biotin-conjugated antiHuman VEGF-A antibody is added and binds to Human VEGF-A captured by the first antibody. Following incubation unbound biotin-conjugated antiHuman VEGF-A antibody is removed during a wash step. Streptavidin-HRP is added and binds to the biotinconjugated anti-Human VEGF-A antibody. Third Incubation - Following incubation unbound Streptavidin-HRP is removed during a wash step, and substrate solution reactive with HRP is added to the wells. Fourth Incubation - A colored product is formed in proportion to the amount of human VEGF-A present in the sample or standard. The reaction is terminated by addition of acid and absorbance is measured at 450 nm. A standard curve is prepared from 7 human VEGF-A standard dilutions and Human VEGF-A sample concentration determined.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Caution: This is intended for research use only.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty