Would you like to log in again?
VEGF-C Human ELISA, BioVendor
Supplier: BioVendor
RAF138R
102552-664EA 1054.98 USD InStock
102552-664
VEGF-C Human ELISA, BioVendor
Assays ELISAs
VEGF-C regulates the lymphatic endothelial cells via their receptor VEGFR3, thus acting as mitogens for these cells.
  • Calibration Range: 0.23-15 pg/mL
  • Limit of Detection: 0.057 pg/mL
  • Cell culture supernatant, Plasma, Serum: 20 µL/well
Vascular endothelial growth factor-C (VEGF-C) is a member of the VEGF family of cytokines. It has been shown to exhibit angiogenic and lymphangiogenic actions. The VEGF family of growth factors and receptors is involved in the development and growth of the vascular endothelial system. Two of its family members, VEGF-C and VEGF-D, regulate the lymphatic endothelial cells via their receptor VEGFR3, thus acting as mitogens for these cells. VEGF-C expression is associated with hematological malignancies. Like VEGF it acts as survival factor on leukemia. Together with the expression of their receptors, VEGF and VEGF-C result in the generation of autocrine loops that may support cancer cell survival and proliferation. Further VEGF-C expression has been shown in gastrointestinal tract malignancies where it correlates with lymphatic invasion, lymphnode metastasis and reduced survival.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty