Would you like to log in again?
Angiopoietin-Like Protein 3, ANGPTL 3, Mouse/Rat ELISA, BioVendor
Supplier: BioVendor
RAG011R
102552-694EA 996.77 USD InStock
102552-694
Angiopoietin-Like Protein 3, ANGPTL 3, Mouse/Rat ELISA, BioVendor
Assays ELISAs
ANGPTL3 is structurally similar to angiopoietins, which are vascular endothelial growth factors.
  • Calibration Range: 0.16-1 ng/mL
  • Limit of Detection: 15 pg/mL
  • Cell culture supernatant, Plasma, Serum: sample diluiton will need to be determined, start at 1:4,000 for mouse and 1:400 for rat samples
Angiopoietin-like proteins ANGPTL3 and ANGPTL4 are secreted proteins mainly expressed in liver that have been demonstrated to regulate triglyceride metabolism by inhibiting the lipolysis of triglyceride-rich lipoproteins. ANGPTL3 is structurally similar to angiopoietins, which are vascular endothelial growth factors. The experimental results show that Angptl3 and Angptl4 function to regulate circulating triglyceride levels during different nutritional states and therefore play a role in lipid metabolism during feeding/fasting through differential inhibition of Lipoprotein lipase (LPL).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty