Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Lcat (from HEK293 cells)

Supplier: BioVendor
RD172122100HEK
102552-810EA 583.35 USD InStock
102552-810
Human Lcat (from HEK293 cells)
Proteins and Peptides
Human Lecithin: cholesterol acyltransferase (LCAT) is a glycoprotein with a molecular mass of approximately 58 kDa. It is the key enzyme responsible for esterification of free cholesterol to cholesteryl esters in circulating plasma lipoproteins, primarily in high density lipoprotein (HDL). The tertiary structure of LCAT is maintained by two disulfide bridges, similar to lipases and other proteins of the α/β hydrolase fold family. Mature LCAT protein is synthesized from a 440 residue precursor by following cleavage of a 24 residue signal peptide. The mature protein contains 416 amino acids and is heavily N-glycosylated. LCAT is abundant in blood plasma and it is present in other organs, including liver, brain and testes. In plasma LCAT is associated with ApoD which frequently co-purify.

Sequence: HVDYKDDDDK PAGFWLLNVL FPPHTTPKAE LSNHTRPVIL VPGCLGNQLE AKLDKPDVVN WMCYRKTEDF FTIWLDLNMF LPLGVDCWID NTRVVYNRSS GLVSNAPGVQ IRVPGFGKTY SVEYLDSSKL AGYLHTLVQN LVNNGYVRDE TVRAAPYDWR LEPGQQEEYY RKLAGLVEEM HAAYGKPVFL IGHSLGCLHL LYFLLRQPQA WKDRFIDGFI SLGAPWGGSI KPMLVLASGD NQGIPIMSSI KLKEEQRITT TSPWMFPSRM AWPEDHVFIS TPSFNYTGRD FQRFFADLHF EEGWYMWLQS RDLLAGLPAP GVEVYCLYGV GLPTPRTYIY DHGFPYTDPV GVLYEDGDDT VATRSTELCG LWQGRQPQPV HLLPLHGIQH LNMVFSNLTL EHINAILLGA YRQGPPASPT ASPEPPPPE

Other names: LCAT, Phosphatidylcholine-sterol acyltransferase, Phospholipid-cholesterol acyltransferase

Ordering information: For research use only.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty