Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GDF7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-11461R-A647
10256-040EA 811.1 USD InStock
10256-040
Anti-GDF7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Growth/differentiation factors (GDFs) are members of the TGF superfamily (1,2). Members of the TGF superfamily are involved in embryonic development and adult tissue homeostasis (1). GDF-1 expression is almost exclusively restricted to the central nervous system and mediates cell differentiation events during embryonic development (3). Neither GDF-3 (Vgr-2) nor GDF-9 contains the conserved cysteine residue which is found in most other TGF superfamily members. GDF-3 is detectable in bone marrow, spleen, thymus and adipose tissue, whereas GDF-9 has only been detected in ovary (4). GDF-5 (also designated CDMP-1) has been shown to induce activation of plasminogen activator, thereby inducing angiogenesis. It is predominantly expressed in long bones during fetal embryonic development and is involved in bone formation. (5). GDF-5 mutations have been identified in mice with the mutation brachypodism (bp), a mutation which affects the length and number of bones in limbs (6). GDF-6 and GDF-7 are closely related to GDF-5 (6). GDF-8 has been shown to be a negative regulator of skeletal muscle mass (1).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GDF7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty