Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NRAS Rabbit Polyclonal Antibody

Supplier: Bioss

BS-1146R
10257-356EA 649.69 USD InStock
10257-356
Anti-NRAS Rabbit Polyclonal Antibody
Antibodies
N-RAS (neuroblastoma RAS viral oncogene homolog) analogously to other GTP-binding proteins (such as Translation Elongation Factor EFTu or signal transducing G-Proteins) RAS proteins are involved in signal transduction pathways, ras gene family is part of the ras superfamily including the mammalian RAS, RAL, RAC, RHO, RAP, and RAB gene families and the yeast homologs like SEC4 and YPT1 genes; genes encode small monomeric proteins of low molecular mass (20-30 kDa) which share at least 30% homology to RAS proteins.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: NRAS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty