Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZNF312B Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-11498R-A555
10258-026EA 811.1 USD InStock
10258-026
Anti-ZNF312B Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Olfactory sensory neurons contain olfactory receptors, which are G protein-coupled receptor proteins that localize to the cilia and display affinity for and bind to a variety of odor molecules. Olfactory neurons send their axons from the olfactory epithelium to the olfactory bulb, which is covered by the CNS basal lamina. FEZF1 (Fez family zinc finger protein 1), also known as Forebrain Embryonic Zinc Finger and Zinc finger protein 312B, is a 475 amino acid nuclear protein that is expressed in the olfactory epithelium and hypothalamus of mice. In FEZF1-deficient mice, axons of olfactory neurons do not reach the olfactory bulb, suggesting that FEXF1 is required for the penetration of olfactory axons though the basal lamina before innervation of the olfactory bulb. When FEZF1 translocates to the nucleus, it induces KRAS overexpression, resulting in activation of ERK-signaling. Overexpression of FEZF1 leads to accelerated proliferation in cultured cells and increased tumor mass in mice. There are three isoforms of FEZF1 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF312B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty