Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) ≥98% (by HPLC)

Supplier: CHEM IMPEX INTL INC MS
Synonyms: Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate

00095-5G 00095-250MG 00095-1G
102614-644EA 29.52 USD InStock
102614-644 102587-074 102587-072
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) ≥98% (by HPLC)
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2)


Formula: C₉H₁₄N₃Na₂O₁₄P₃
MW: 527.12 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078193
CAS Number: 36051-68-0
Order Now
Specification Test Results
Appearance White powder
Solubility Clear colorless solution (50 mg/mL in Water)
Water Content (Karl Fisher) ≤12.0%
UV Ratios, A250/A260 at pH 7.0 0.81 - 0.87
UV Ratios, A280/A260 at pH 7.0 0.94 - 1.00
Purity (HPLC) ≥98%
Purity (UV) ≥96% (on Dry Basis)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty