Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gentamicin sulfate USP

Supplier: 0000014015
Synonyms: NMD Inhibitor IIGaramycinGentiomycin CGentamicin sulfate saltGentamicin sulphate salt

00149-5G 00149-1KG 00149-25G 00149-100G 00149-250G
102562-124EA 2440.77 USD InStock
102562-124 102606-330 102614-746 102586-426 102614-744
Gentamicin sulfate USP
Gentamicin sulphate
White to off-white powder

  • Optical rotation: +107° to +121°
  • Potency: ≥ 590 µg/mg (Anhydrous basis)


Melting Pt: 218…237 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS Number: 1405-41-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty