Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-LRP5L Rabbit Polyclonal Antibody

Supplier: Bioss

BS-11590R
10262-828EA 649.69 USD InStock
10262-828
Anti-LRP5L Rabbit Polyclonal Antibody
Antibodies
Members of the LDL receptor gene family, including LDLR (low density lipo-protein receptor), LRPs (low density lipoprotein related proteins), Megalin (also designated GP330), VLDLR (very low density lipoprotein receptor) and ApoER2, are characterized by a cluster of cysteine-rich class A repeats, epidermal growth factor (EGF)-like repeats, YWTD repeats and an O-linked sugar domain. Of the known family members, LRP5 is most closely related to LRP1. However, LRP5 has a unique organization of EGF and LDLR repeats compared to other LDLR family members and likely represents a new category in this family. LRP is expressed in rat tibia, specifically in areas of the bone that are involved in remodeling. LRP5 is a Wnt coreceptor that binds to Axin and regulates the canonical Wnt signaling pathway. LRP5 affects bone mass accrual during growth. Mutations in LRP5 cause the autosomal recessive disorder osteoporosis-pseudoglioma syndrome (OPPG). LRP5L is a related protein that exists as two isoforms produced by alternative splicing.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRP5L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty