Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dess-Martin periodinane ≥95%, Powder

Supplier: CHEM IMPEX INTL INC MS
Synonyms: Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl esterDMP1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one(1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one

14504-1KG 14504-5G 14504-100G 14504-25G 14504-1G 14504-250G
102554-764EA 3762.52 USD InStock
102554-764 102632-994 102578-046 102621-558 102578-042 102578-044
Dess-Martin periodinane ≥95%, Powder
Dess-Martin periodinane


Formula: C₁₃H₁₃IO₈
MW: 424.15 g/mol
MDL Number: MFCD00130127
CAS Number: 87413-09-0
Order Now
Specification Test Results
Molecular Formula C13H13IO8
Molecular Weight 424.14
Melting Point 128-135 °C
MDL Number MFCD00130127
Storage Temperature Store at 0-8°C
Color White to Off-White

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty