Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EBF2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-11740R-CY7
10266-322EA 811.1 USD InStock
10266-322
Anti-EBF2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Early B-cell factor 2 is a 575 amino acid protein belonging to the COE family of proteins, whose members are all helix-loop-helix transcription factors. EBF2 is a transcription factor which, in synergy with the Wnt-responsive LEF1/CTNNB1 pathway, activates the decoy receptor for RANKL, OPG, in osteoblasts. OPG, in turn, regulates osteoclast differentiation. Lack of EBF2 has been found to cause a small defect in the terminal differentiation of osteoblasts, along with reduced bone mass and an increase in osteoclasts. Localized to the nucleus, EBF2 forms a homodimer or a heterodimer with a related family member.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EBF2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty