Would you like to log in again?
Protein Loading Buffer, 4X, Li-Cor
Supplier: Li-Cor
928-40004
102673-500EA 51.69 USD InStock
102673-500
Protein Loading Buffer, 4X, Li-Cor
Buffers Electrophoresis Buffers Loading Buffers
Protein loading buffer (4X) is used as a loading buffer for protein gel electrophoresis.
This buffer is optimized for use with SDS-page and Tris-glycine-SDS running buffer.
It contains Tris-HCl (pH 6.8) 125 mM, glycerol 50%, SDS 4%, orange G 0.2% (w/v).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty