Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
DiATOME® Ultra and Cryo Diamond Knife for AFM, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
30-AFM-UDL 30-AFM-UDR 30-AFM-CDL 20-AFM-CDL 20-AFM-UDL 30-AFM-CDR 20-AFM-CDR 20-AFM-UDR
102680-568EA 5962.47 USD InStock
102680-568 102680-698 102680-696 102680-574 102680-694 102680-572 102680-692 102680-570
DiATOME® Ultra and Cryo Diamond Knife for AFM, Electron Microscopy Sciences
Knives
These high quality Diatome diamond knives are not just perfect for producing ultrathin and semi-thin sections, but also for surfacing sectioning of all kinds of biological and industrial specimens.
  • Extremely smooth sample surfaces and best possible structure preservation
  • Available in 35° with a cutting range of 10-100 nm in 2 and 3 mm sizes
Instead of a section as for TEM, the specimen surface is investigated using AFM. In order to achieve the best results for AFM investigation, only the highest quality diamond knives should be used. Diatome ultra AFM and cryo AFM knives are specially tested to ensure that they meet the increased quality requirements of AFM investigation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty