Would you like to log in again?
DiATOME® Cryo Diamond Knives, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
27-CDSE 35-CDSR 30-CDSR 24-CWLE 27-CDLR 30-CWLE 30-CWSE 24-CDLR 30-CWL 15-CDSE 30-CD25 21-CWSR 18-CDL 24-CDSE 30-CDSE 21-CWLE 24-CWSR 35-CDLR 30-CDL 15-CDLE 35-CWLE 18-CDS 21-CWSE 40-CWLE 40-CWSE 35-CWLR 24-CWLR 30-CDLR 18-CWL 35-CWSR 40-CWSR 27-CWSE 18-CDLR 40-CDS 18-CDSE 18-CWSE 27-CDLE 18-CDLE 27-CWLR 24-CWL 40-CDSR 21-CDLR 15-CDSR 21-CWLR 40-CDL 15-CDLR 15-CWS 15-CWLR 21-CWL 35-CDS 35-CDL 18-CDSR 18-CWLR 27-CDSR 35-CWSE 40-CWS 21-CWS 30-CWSR 27-CWSR 18-CWLE 15-CDL 30-CD25R 30-CWLR 15-CWSR 18-CWS 21-CDL 24-CWSE 27-CDL 21-CDS 30-CDS 30-CDLE 18-CWSR 21-CDLE 27-CWS 30-CWS 24-CDLE 21-CDSR 27-CDS 21-CDSE 27-CWL 15-CDS 40-CDLR 35-CWS 24-CDS 40-CWL 15-CWL 24-CWS 40-CDLE 24-CDSR 35-CDLE 15-CWLE 35-CWL 27-CWLE 40-CWLR 15-CWSE 24-CDL 40-CDSE 35-CDSE
102680-712EA 5909.26 USD InStock
102680-712 102680-678 102680-710 102680-798 102680-676 102680-796 102680-554 102680-674 102680-552 102680-794 102680-718 102680-716 102680-714 102680-672 102680-550 102680-792 102680-670 102680-790 102680-548 102680-702 102680-668 102680-700 102680-546 102680-666 102680-544 102680-664 102680-542 102680-662 102680-708 102680-706 102680-704 102680-584 102680-724 102680-602 102680-722 102680-600 102680-720 102680-606 102680-726 102680-604 102680-760 102680-636 102680-514 102680-756 102680-634 102680-512 102680-754 102680-632 102680-598 102680-510 102680-630 102680-596 102680-518 102680-638 102680-758 102680-516 102680-594 102680-592 102680-590 102680-504 102680-624 102680-502 102680-622 102680-588 102680-500 102680-620 102680-586 102680-508 102680-628 102680-506 102680-626 102680-540 102680-660 102680-812 102680-658 102680-536 102680-810 102680-656 102680-776 102680-534 102680-532 102680-774 102680-498 102680-772 102680-538 102680-496 102680-770 102680-768 102680-800 102680-766 102680-764 102680-642 102680-762 102680-640 102680-808 102680-806 102680-804 102680-802
DiATOME® Cryo Diamond Knives, Electron Microscopy Sciences
Knives
Diatome cryo diamond knives may be used for all low temperature situations.
  • Produce thinner sections
  • Ultrathin and semithin sections with the same knife
  • Easy to produce long ribbons of sections
  • Less compression and greater uniformity of sections
  • No waiting for temperature equilibration (no knife manufacturing or knife changing)
It is used in sectioning of cryo-protected specimens, frozen hydrated specimens and industrial samples such as polymers and rubber. Section thickness range 30 nm – 1 µm.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty