Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Asbestos TEM Cassette 25 mm, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
65216-50
102681-078PK 112.34 USD InStock
102681-078
Asbestos TEM Cassette 25 mm, Electron Microscopy Sciences
Filters Cassette Filters
This PCM 25 mm air sampling cassettes are designed and manufactured to conform to all NIOSH, OSHA, and EPA standards. The cassette housings are injection molded in-house and are individually assembled and preloaded under stringent quality control standards.
  • Quality 0.45 µm MCE filter material
  • 5.0 µm Filter is placed under the 0.45 µm filter as a diffuser
  • 2" Static conductive extension cowl
  • Meets NIOSH 7402 specifications
  • Meets AEHRA requirements
  • Preloaded – ready-to-use
Once assembled, each cassette is individually tested prior to packaging for shipment. Random lot sample testing is performed daily by an independent laboratory to ensure low background fiber counts. Laboratory results are available upon request.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty