Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cassette, PC Filters, 37 mm, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
65220-24 65220-38 65220-28 65220-34
102681-100PK 234.85 USD InStock
102681-100 102681-096 102681-098 102681-094
Cassette, PC Filters, 37 mm, Electron Microscopy Sciences
Filters Cassette Filters
The 37 mm Air Sampling Cassette with polycarbonate Filters are designed to meet all NIOSH, OSHA, and EPA requirement for air sampling standards
All 37 mm cassette housings are manufactured in house and are available in both two and three piece configurations. All 37 mm cassettes are individually assembled and preloaded under stringent quality assurance standards with only the finest quality membrane and support pads.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty