Would you like to log in again?
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
65240-63 65240-64 65240-60 65240-61
102681-156EA 2267.66 USD InStock
102681-156 102681-158 102681-152 102681-154
mini-BUCK™ Primary Flow Calibrator, Electron Microscopy Sciences
Flow Meters
The mini-BUCK™ Primary Flow Calibrator is the industry standard for day-to-day flow verification and calibration of your personal and environmental sampling pumps.
  • No more lugging oversized calibration panels
  • No more rotameter inaccuracies
It is also suitable for verifying rotameters, setting instrument panels, and calibrating laboratory instruments. The mini-BUCK™ is accurate at any attitude and can be used for flow measurements with sampling media.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty