Would you like to log in again?
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68047-PS4R-NA 68047-PS4R-NP 68047-PS4R 68047-PS4R-GR
102096-140EA 1154.93 USD InStock
102096-140 102096-144 102096-142 102681-172
PS4R Reflected Light Micrometer Scale, Electron Microscopy Sciences
Standards Color Reference and Spectroscopy Standards
Reflected light calibration slide with 0.1" length scale subdivided into 0.001" divisions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty