Would you like to log in again?
Curved Forceps with Medium Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72910-6 72910-6S 72910-7S 72910-7
102098-080EA 59.24 USD InStock
102098-080 102681-216 102681-218 102098-082
Curved Forceps with Medium Tips, Electron Microscopy Sciences
Forceps
EMS Brand, Swiss made forceps from low carbon austenitic steel, corrosion resistance, high quality, curved tips, available with different length and tips style.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty