Would you like to log in again?
Curved Forceps with Fine Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72911-6S 72911-7 72911-6 72911-7S
102098-090EA 52.82 USD InStock
102098-090 102681-222 102098-088 102681-220
Curved Forceps with Fine Tips, Electron Microscopy Sciences
Forceps
EMS Brand, Swiss made forceps from low carbon austenitic steel.
  • High corrosion resistance
  • High quality
  • Curved tips
Available with different length and tips style.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty