Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Multi Pin Holder for 8 Pin Stubs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75952-08H 75952-08
102681-264EA 174.22 USD InStock
102681-264 102098-306
Multi Pin Holder for 8 Pin Stubs, Electron Microscopy Sciences
Specimen Mounting Supplies
This 50 mm in diameter multi pin holder is designed to save time. It is made from machined aluminum with stainless steel Allen wrench screws.
It accommodates up to eight standard 12.7 mm pin stubs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty