Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tungsten Probe Tip, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

75960-02
102681-276PK 339.84 USD InStock
102681-276
Tungsten Probe Tip, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Custom dual taper tip designed from tungsten. Tip radius is 0.5 µm with 8 to 10° taper angle for maximum lifetime service.

  • For use with AutoProbe™ 200
  • Shank diameter is 0.508 mm (0.020")
  • Needle length is 34.036 mm (1.34")

The reduced taper angle allows for extended needle life for those users who cut and reshape their needles in the FIB.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty