Would you like to log in again?
Omniprobe Lift-Out Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75964-02 75964-01
102681-290PK 479.51 USD InStock
102681-290 102681-292
Omniprobe Lift-Out Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Custom copper lift-out grids specifically designed for in-situ lift-out.
  • Multiple indexed mounting locations
  • With vertical bars and “V” shaped attachment surfaces
Measures 3 mm in diameter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty