Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Omniprobe Short-Cut™ Coupons, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
75977-MO 75975-MO 75976-MO 75977-CU 75974-MO 75974-CU 75976-CU 75975-CU
102681-324PK 216.39 USD InStock
102681-324 102681-326 102681-332 102681-320 102681-334 102681-322 102681-330 102681-328
Omniprobe Short-Cut™ Coupons, Electron Microscopy Sciences
Graticules Reticles and Grids
TEM sample grid coupons designed specially for use in the Short-Cut™ tool. Available in two versions: One made from pure copper and the other is molybdenum coated copper.

  • Frontside Thinning, 45°, 45° cut
  • Backside Thinning, 45°, 45° cut
  • Frontside Thinning, 26.5°, 26.5° cut
  • Backside Thinning, 26.5°, 26.5° cut

Frontside Thinning, 26.5°, the center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut 26.5°.

Backside Thinning, 26.5°, the center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut 26.5°.

Frontside Thinning, 45°, the center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut 45°.

Backside Thinning, 45°, the center portion is a standard 3 mm grid into which the sample loaded needle is swaged and cut 45°.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty