Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Denka M3 LaB6 Cathode, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80900-101 80904-40 80906 80904-61 80900-61 80906-11 80906-41 80904-60 80900-60 80908-40 80908-81 80906-20 80900-11 80908-100 80906-10 80908-20 80904-41 80904 80908-11 80904-80 80906-80 80900-40 80914 80908-21 80904-5 80900-20 80908-60 80900-10 80906-100 80900-100 80904-21 80904-10 80906-61 80914-20 80908-61 80906-5 80906-21 80904-20 80904-100 80900-41 80906-60 80908 80908-10 80908-41 80900 80902 80912 80906-40 80900-21 80900-80 80908-80 80910 80900-5 80908-5 80904-81 80906-81 80904-101
102681-446EA 1931.62 USD InStock
102681-446 100492-390 102681-402 100492-392 102681-404 102681-448 100492-394 102681-486 102681-442 102681-444 102681-400 102681-488 100502-146 100502-148 102681-406 100502-144 102681-408 102681-450 102681-412 102681-456 100502-150 102681-458 102681-414 102681-452 102681-410 102681-454 102681-416 102681-418 102680-468 102681-460 102681-462 102681-468 102681-424 102681-426 102681-464 102681-420 102681-466 102681-422 102681-428 102681-394 102681-470 102681-396 102681-472 102681-392 102681-434 102681-478 102681-436 102681-398 102681-474 102681-430 102681-432 102681-476 102681-438 102681-482 102681-440 102681-484 102681-480
Denka M3 LaB6 Cathode, Electron Microscopy Sciences
Lights Hollow Cathode Lamps
Lanthanum Hexaboride is the optimum cathode material for use in SEM, TEM, X-Ray Microanalysis, and Electron Beam Lithography.
  • Low potential barrier (work function)
  • High melting point and stability at high temperatures
  • Low vapor pressure-reduces evaporation lost
  • Chemical stability
LaB6 beam is ten times brighter than tungsten, has superior resolution-wide range of acceleration voltages. It provides sharp and clear pictures even to the smallest details. The initial brightness of a Denka Model 3 cathode (1×106 A/cm2.Str) is substantially better than that of competing mini-vogel type cathodes (typically 5×105 A/cm2.Str).
Standard (Round) Tips are provided with a 90° cone and 15 µm radius round tip. Flat (Truncated) Tips are recommended for applications where stability and long lifetime are the prime considerations, and where some degree of brightness can be sacrificed. Flat tips are available in eight configurations. Sharp tips are recommended for applications where initial brightness is the foremost consideration and where shorter lifetime can be accepted. Sharp tips are available in two configurations. All standard Denka M-3 LaB6 Cathodes are provided with a 90° cone angle and a 15 µm radius round tip. These are just the standard Denka LaB6 Cathodes that are in stock. The complete line of Denka Lab6 filaments is available through Electron Microscopy Sciences.
Heat and vacuum are the final determinants of any cathode tip's life, but at 1550 °C and a vacuum of 10 - 7 Torr, a service life of about 500 hours can be achieved.
Every Denka Model 3 cathode offers a stability of better than 3% per hour at 1550 °C and Denka Model3 LaB6 cathodes are designed as direct replacement for tungsten filaments
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty